accessibility menu, dialog, popup

UserName

CH3OONCO

CAS RN 41221-47-0

IUPAC Name: methyl 3-isocyanatobenzoate
Synonyms: methyl 3-isocyanatobenzoate
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: WBGWGERFPSYHDT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1)N=C=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results