accessibility menu, dialog, popup

UserName

CAS RN 4114-31-2

H3COONHNH2

CAS RN 4114-31-2

IUPAC Name: ethoxycarbohydrazide
Synonyms: hydrazinecarboxylic acid, ethyl ester
Molecular Weight (g/mol): 104.11
Molecular Formula: C3H8N2O2
InChi Key: VYSYZMNJHYOXGN-UHFFFAOYSA-N
SMILES: CCOC(=O)NN
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)