accessibility menu, dialog, popup

UserName

CAS RN 4107-98-6

CH3H3CNCH3CH3

CAS RN 4107-98-6

IUPAC Name: N,N-bis(propan-2-yl)aniline
Synonyms: N,N-diisopropylaniline
Molecular Weight (g/mol): 177.29
Molecular Formula: C12H19N
InChi Key: OVSARSKQWCLSJT-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)