accessibility menu, dialog, popup

UserName

CAS RN 41024-91-3

H3CSOOOOOOOOSOOCH3

CAS RN 41024-91-3

IUPAC Name: 14-[(4-methylbenzenesulfonyl)oxy]-3,6,9,12-tetraoxatetradecan-1-yl 4-methylbenzene-1-sulfonate
Synonyms: 14-[(4-methylbenzenesulfonyl)oxy]-3,6,9,12-tetraoxatetradecan-1-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 546.65
Molecular Formula: C24H34O10S2
InChi Key: BUHGDYPBQWWWQS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOCCOS(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results