accessibility menu, dialog, popup

UserName

CAS RN 40987-25-5

ONCl

CAS RN 40987-25-5

IUPAC Name: 4-benzyl-2-(chloromethyl)morpholine
Synonyms: 4-benzyl-2-(chloromethyl)morpholine
Molecular Weight (g/mol): 225.72
Molecular Formula: C12H16ClNO
InChi Key: GVWRZZNYCOTWNN-UHFFFAOYNA-N
SMILES: ClCC1CN(CC2=CC=CC=C2)CCO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results