accessibility menu, dialog, popup

UserName

CAS RN 4097-49-8

CH3H3CCH3OHNOONOO

CAS RN 4097-49-8

IUPAC Name: 4-tert-butyl-2,6-dinitrophenol
Synonyms: 4-tert-butyl-2,6-dinitrophenol
Molecular Weight (g/mol): 240.22
Molecular Formula: C10H12N2O5
InChi Key: NJBDTWSOYUZQPM-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O

 No Refinements