CAS RN 407640-40-8
IUPAC Name:
2-(4-fluoro-2-methylphenyl)acetic acid
Synonyms:
(4-fluoro-2-methylphenyl)acetic acid
Molecular Weight (g/mol):
168.17
Molecular Formula:
C9H9FO2
InChi Key:
KOZXQTAPRQFRLT-UHFFFAOYSA-N
SMILES:
CC1=C(CC(O)=O)C=CC(F)=C1
1
–
1
of
1
results
1
4-Fluoro-2-methylphenylacetic acid, 97%, Thermo Scientific™
CAS: 407640-40-8 Molecular Formula: C9H9FO2 Molecular Weight (g/mol): 168.167 MDL Number: MFCD00049318 InChI Key: KOZXQTAPRQFRLT-UHFFFAOYSA-N Synonym: 4-fluoro-2-methylphenylacetic acid,2-4-fluoro-2-methylphenyl acetic acid,4-fluoro-2-methylphenyl acetic acid,4-fluoro-2-methyl-phenyl-acetic acid,benzeneacetic acid, 4-fluoro-2-methyl,pubchem4174,2-4-fluoro-2-methyl-phenyl acetic acid,acmc-1akub,timtec-bb sbb027289,4-fluoro-2-methylphenylaceticacid PubChem CID: 5016950 IUPAC Name: 2-(4-fluoro-2-methylphenyl)acetic acid SMILES: CC1=C(C=CC(=C1)F)CC(=O)O