accessibility menu, dialog, popup

UserName

CAS RN 40733-28-6

(S)(R)O(R)NHNCH3OSiOOHOH3CCH3H3CCH3H3C

CAS RN 40733-28-6

IUPAC Name: 6-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}pyridine-3-carboxylic acid
Synonyms: 6-[4-(tert-butoxycarbonyl)piperazin-1-yl]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 307.35
Molecular Formula: C15H21N3O4
InChi Key: PWYGTZUOLAGDNK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=N1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results