accessibility menu, dialog, popup

UserName

NClClOHOCl

CAS RN 406676-18-4

IUPAC Name: 2,3,5-trichloropyridine-4-carboxylic acid
Synonyms: 2,3,5-trichloropyridine-4-carboxylic acid
Molecular Weight (g/mol): 226.44
Molecular Formula: C6H2Cl3NO2
InChi Key: TYMTYHMQJHSRIF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C(Cl)=NC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results