accessibility menu, dialog, popup

UserName

OOFFF

CAS RN 406-93-9

IUPAC Name: 4,4,4-trifluorobutanoate
Synonyms: 4,4,4-trifluorobutanoate
Molecular Weight (g/mol): 141.07
Molecular Formula: C4H4F3O2
InChi Key: WTUCTMYLCMVYEX-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results