accessibility menu, dialog, popup

UserName

CAS RN 40365-61-5

HCOO

CAS RN 40365-61-5

IUPAC Name: 2-(but-3-yn-1-yloxy)oxane
Synonyms: 2-(but-3-yn-1-yloxy)oxane
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: ZQZSNKJFOFAJQX-UHFFFAOYNA-N
SMILES: C#CCCOC1CCCCO1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)