accessibility menu, dialog, popup

UserName

CAS RN 403-20-3

H3COFOO

CAS RN 403-20-3

IUPAC Name: 3-fluoro-4-methoxybenzoate
Synonyms: 3-fluoro-4-methoxybenzoate
Molecular Weight (g/mol): 169.13
Molecular Formula: C8H6FO3
InChi Key: HYNNNQDQEORWEU-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1F)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results