accessibility menu, dialog, popup

UserName

H2NNHNN

CAS RN 4023-02-3

IUPAC Name: 1H-pyrazole-1-carboximidamide
Synonyms: pyrazole-1-carboximidamide
Molecular Weight (g/mol): 110.12
Molecular Formula: C4H6N4
InChi Key: UCQFSGCWHRTMGG-UHFFFAOYSA-N
SMILES: NC(=N)N1C=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)