accessibility menu, dialog, popup

UserName

H3COONH2NH2HClHCl

CAS RN 40216-82-8

IUPAC Name: methyl 2,5-diaminopentanoate dihydrochloride
Synonyms: methyl 2,5-diaminopentanoate dihydrochloride
Molecular Weight (g/mol): 219.11
Molecular Formula: C6H16Cl2N2O2
InChi Key: SNFZNVWAQDVFAK-UHFFFAOYNA-N
SMILES: Cl.Cl.COC(=O)C(N)CCCN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results