accessibility menu, dialog, popup

UserName

FFFF

CAS RN 402-44-8

IUPAC Name: 1-fluoro-4-(trifluoromethyl)benzene
Synonyms: 1-fluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 164.1
Molecular Formula: C7H4F4
InChi Key: UNNNAIWPDLRVRN-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)