accessibility menu, dialog, popup

UserName

CAS RN 401892-83-9

H3COSFFFFF

CAS RN 401892-83-9

IUPAC Name: 1-[4-(pentafluoro-λ⁶-sulfanyl)phenyl]ethan-1-one
Synonyms: 1-[4-(pentafluoro-λ⁶-sulfanyl)phenyl]ethanone
Molecular Weight (g/mol): 246.2
Molecular Formula: C8H7F5OS
InChi Key: BKEQOPIQGDSWEQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results