accessibility menu, dialog, popup

UserName

CAS RN 40187-51-7

H3COOHH2NO

CAS RN 40187-51-7

IUPAC Name: 5-acetyl-2-hydroxybenzamide
Synonyms: benzamide, 5-acetyl-2-hydroxy-
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: LWAQTCWTCCNHJR-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(O)C(=C1)C(N)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)