CAS RN 40165-87-5
CAS RN 40165-87-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxynonanoic Acid, TRC
CAS: 40165-87-5 Molecular Formula: C9 H18 O3 Molecular Weight (g/mol): 174.24 Synonym: (+/-)-3-Hydroxynonanoic Acid,3-Hydroxynonanoic Acid,β-Hydroxynonanoic Acid IUPAC Name: 3-hydroxynonanoic acid SMILES: CCCCCCC(O)CC(=O)O