accessibility menu, dialog, popup

UserName

CH3OONCl

CAS RN 40134-18-7

IUPAC Name: methyl 2-chloropyridine-3-carboxylate
Synonyms: methyl 2-chloropyridine-3-carboxylate
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: MYGAJZBZLONIBZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CN=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results