accessibility menu, dialog, popup

UserName

CAS RN 40101-17-5

CH3OOOOH3C

CAS RN 40101-17-5

IUPAC Name: bis(3-methoxyphenyl)ethane-1,2-dione
Synonyms: bis(3-methoxyphenyl)ethane-1,2-dione
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: PJGXOGKIVAJFTE-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1)C(=O)C(=O)C1=CC(OC)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results