accessibility menu, dialog, popup

UserName

NH2NOOFFF

CAS RN 401-94-5

IUPAC Name: 3-nitro-5-(trifluoromethyl)aniline
Synonyms: 3-nitro-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 206.12
Molecular Formula: C7H5F3N2O2
InChi Key: LTVWXWWSCLXXAT-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1)C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results