CAS RN 4008-48-4
CAS RN 4008-48-4
IUPAC Name:
5-nitroquinolin-8-ol
Synonyms:
nitroxoline nicene forte noxibiol 5-nitroxine 5-nok nitroxolina [inn-spanish] 5-nitrox nitroxoline (inn) and its salts nitroxolinum 5-nitroks
Molecular Weight (g/mol):
190.16
Molecular Formula:
C9H6N2O3
InChi Key:
RJIWZDNTCBHXAL-UHFFFAOYSA-N
SMILES:
OC1=C2N=CC=CC2=C(C=C1)[N+]([O-])=O
1
–
3
of
3
results
1
Nitroxoline, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Nitroxoline, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Nitroxoline, MedChemExpress
MedChemExpress Nitroxoline is an antibiotic that has proven to be very effective at combating biofilm infections.