accessibility menu, dialog, popup

UserName

CAS RN 400771-41-7

IUPAC Name: 1-[3-chloro-5-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[3-chloro-5-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 209.6
Molecular Formula: C8H7ClF3N
InChi Key: GCLAXYMPBCHQIK-UHFFFAOYSA-N
SMILES: NCC1=CC(=CC(Cl)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results