accessibility menu, dialog, popup

UserName

CAS RN 40064-34-4

IUPAC Name: hydrogen piperidin-4-one hydrate chloride
Synonyms: hydrogen 4-piperidinone hydrate chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: SXWRTZOXMUOJER-UHFFFAOYSA-N
SMILES: [H+].O.[Cl-].O=C1CCNCC1
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results