accessibility menu, dialog, popup

UserName

CAS RN 400-38-4

H3CCH3OOFFF

CAS RN 400-38-4

IUPAC Name: propan-2-yl 2,2,2-trifluoroacetate
Synonyms: isopropyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H7F3O2
InChi Key: ASAXRKSDVDALDT-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)