accessibility menu, dialog, popup

UserName

CAS RN 39994-75-7

IUPAC Name: (2S,3R)-3-hydroxy-1-methoxy-1-oxobutan-2-aminium
Synonyms: (2S,3R)-3-hydroxy-1-methoxy-1-oxobutan-2-aminium
Molecular Weight (g/mol): 134.15
Molecular Formula: C5H12NO3
InChi Key: TVHCXXXXQNWQLP-DMTCNVIQSA-O
SMILES: COC(=O)[C@@H]([NH3+])[C@@H](C)O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)