accessibility menu, dialog, popup

UserName

CAS RN 3992-42-5

NN(R)O(R)(S)ONH2HOHOHCl

CAS RN 3992-42-5

IUPAC Name: 4-amino-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloridyl
Synonyms: deoxycytidine hydrochloridyl
Molecular Weight (g/mol): 262.67
Molecular Formula: C9H13ClN3O4
InChi Key: RGRYRUDWHSHWGG-UHFFFAOYNA-N
SMILES: [Cl].NC1=NC(=O)N(C=C1)C1CC(O)C(CO)O1
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)