accessibility menu, dialog, popup

UserName

CAS RN 39906-04-2

CH3NClNH2ClN

CAS RN 39906-04-2

IUPAC Name: 4,6-dichloro-2-methylpyrimidin-5-amine
Synonyms: 4,6-dichloro-2-methylpyrimidin-5-amine
Molecular Weight (g/mol): 178.02
Molecular Formula: C5H5Cl2N3
InChi Key: FKRXXAMAHOGYNT-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=C(N)C(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results