accessibility menu, dialog, popup

UserName

CAS RN 39811-14-8

ClHNNOHO

CAS RN 39811-14-8

IUPAC Name: 6-chloro-1H-1,3-benzodiazole-2-carboxylic acid
Synonyms: 5-chloro-3H-1,3-benzodiazole-2-carboxylic acid
Molecular Weight (g/mol): 196.59
Molecular Formula: C8H5ClN2O2
InChi Key: NZIHMSYSZRFUQJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC2=CC=C(Cl)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results