accessibility menu, dialog, popup

UserName

NOOHNBr

CAS RN 397864-11-8

IUPAC Name: 3-bromo-7-nitro-1H-indole
Synonyms: 3-bromo-7-nitro-1H-indole
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: ZJCDMQUXOJHHBJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2NC=C(Br)C2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results