accessibility menu, dialog, popup

UserName

CAS RN 3972-65-4

CH3H3CCH3Br

CAS RN 3972-65-4

IUPAC Name: 1-bromo-4-tert-butylbenzene
Synonyms: 4-tert-butyl-1-bromobenzene
Molecular Weight (g/mol): 213.12
Molecular Formula: C10H13Br
InChi Key: XHCAGOVGSDHHNP-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(Br)C=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)