CAS RN 39718-07-5
CAS RN 39718-07-5
IUPAC Name:
5-methylbenzene-1,3-dicarbonitrile
Synonyms:
5-methylbenzene-1,3-dicarbonitrile
Molecular Weight (g/mol):
142.16
Molecular Formula:
C9H6N2
InChi Key:
LYSGVAGOLJKEGK-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC(=C1)C#N)C#N