accessibility menu, dialog, popup

UserName

CAS RN 3970-21-6

IUPAC Name: 1-(chloromethoxy)-2-methoxyethane
Synonyms: 1-(chloromethoxy)-2-methoxyethane
Molecular Weight (g/mol): 124.56
Molecular Formula: C4H9ClO2
InChi Key: BIAAQBNMRITRDV-UHFFFAOYSA-N
SMILES: COCCOCCl
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)