accessibility menu, dialog, popup

UserName

CAS RN 39684-80-5

IUPAC Name: tert-butyl N-(2-bromoethyl)carbamate
Synonyms: tert-butyl N-(2-bromoethyl)carbamate
Molecular Weight (g/mol): 224.1
Molecular Formula: C7H14BrNO2
InChi Key: TZRQZPMQUXEZMC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCBr
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)