accessibility menu, dialog, popup

UserName

CH3OHCl

CAS RN 39652-32-9

IUPAC Name: (3-chloro-4-methylphenyl)methanol
Synonyms: (3-chloro-4-methylphenyl)methanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: NLRJZTGNCBMNKS-UHFFFAOYSA-N
SMILES: CC1=CC=C(CO)C=C1Cl
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results