CAS RN 39603-24-2
CAS RN 39603-24-2
IUPAC Name:
5,7-dimethyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms:
5,7-dimethyl-1H-indole-2,3-dione
Molecular Weight (g/mol):
175.19
Molecular Formula:
C10H9NO2
InChi Key:
HFZSCCJTJGWTDZ-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C2NC(=O)C(=O)C2=C1
1
–
1
of
1
results
1
5,7-Dimethylisatin, 96%
CAS: 39603-24-2 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.187 MDL Number: MFCD00047219 InChI Key: HFZSCCJTJGWTDZ-UHFFFAOYSA-N Synonym: 5,7-dimethylisatin,5,7-dimethylindoline-2,3-dione,5,7-dimethyl-indole-2,3-dione,5,7-dimethyl-2,3-dihydro-1h-indole-2,3-dione,1h-indole-2,3-dione, 5,7-dimethyl,indole-2,3-dione, 5,7-dimethyl,1h-indole-2,3-dione, 5,7-dimethyl-9ci,acmc-20a4zq,isatin-based compound, 48,1h-indole-2,3-dione,5,7-dimethyl PubChem CID: 38296 IUPAC Name: 5,7-dimethyl-1H-indole-2,3-dione SMILES: CC1=CC(=C2C(=C1)C(=O)C(=O)N2)C