accessibility menu, dialog, popup

UserName

CAS RN 39576-82-4

H3CNNClCl

CAS RN 39576-82-4

IUPAC Name: 2,4-dichloro-6-methylquinazoline
Synonyms: 2,4-dichloro-6-methylquinazoline
Molecular Weight (g/mol): 213.06
Molecular Formula: C9H6Cl2N2
InChi Key: VZDVVLXYAOVNRW-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=C(Cl)N=C(Cl)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results