accessibility menu, dialog, popup

UserName

CAS RN 39512-49-7

HClHONHCl

CAS RN 39512-49-7

IUPAC Name: hydrogen 4-(4-chlorophenyl)piperidin-4-ol chloride
Synonyms: hydrogen 4-(4-chlorophenyl)piperidin-4-ol chloride
Molecular Weight (g/mol): 248.15
Molecular Formula: C11H15Cl2NO
InChi Key: PXAJMBHRLGKPQV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (8)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)