CAS RN 394-25-2
IUPAC Name:
1-methoxy-2-nitro-4-(trifluoromethyl)benzene
Synonyms:
1-methoxy-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
221.14
Molecular Formula:
C8H6F3NO3
InChi Key:
MAAFHLOZHBKYTG-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F
1
–
1
of
1
results
1
2-Nitro-4-(trifluoromethyl)anisole, 98%, Thermo Scientific™
CAS: 394-25-2 Molecular Formula: C8H6F3NO3 Molecular Weight (g/mol): 221.135 MDL Number: MFCD00007099 InChI Key: MAAFHLOZHBKYTG-UHFFFAOYSA-N Synonym: 4-methoxy-3-nitrobenzotrifluoride,1-methoxy-2-nitro-4-trifluoromethyl benzene,2-nitro-4-trifluoromethyl anisole,2-nitro-4-trifluoromethylanisole,3-nitro-4-methoxybenzotrifluoride,4-trifluoromethyl-2-nitroanisole,benzene, 1-methoxy-2-nitro-4-trifluoromethyl,4-trifluoromethyl-1-methoxy-2-nitrobenzene,pubchem13493,4-methoxy-3-nitrobezotrifluoride PubChem CID: 96748 IUPAC Name: 1-methoxy-2-nitro-4-(trifluoromethyl)benzene SMILES: COC1=C(C=C(C=C1)C(F)(F)F)[N+](=O)[O-]