CAS RN 39365-88-3
IUPAC Name:
tetrapotassium 1-oxo-1λ⁶-disulfene-1,1-bis(olate) sulfanide
Synonyms:
tetrapotassium Hypo sulfanide
Molecular Weight (g/mol):
301.58
Molecular Formula:
HK4O3S3
InChi Key:
LBOBJZFANBWPPF-UHFFFAOYSA-K
SMILES:
[SH-].[K+].[K+].[K+].[K+].[O-]S([O-])(=O)=S
1
–
2
of
2
results
1
Potash, sulfurated, S 11.8% min as sulfide
CAS: 39365-88-3 Molecular Formula: K4O3S3 Molecular Weight (g/mol): 300.57 MDL Number: MFCD00064604 InChI Key: YFKFGPGTVQHNEF-UHFFFAOYSA-L Synonym: potash, sulfurated,sulfurated potash,potash, sulfurated usp,liver of sulfur,2k.k2s.so32,potash sulfurated liver of sulfur,dipotassium hypo potassium sulfide,potash, sulfurated, s as sulfide,dipotassium thiosulfate mixture with potassium sulfide,dipotassium thiosulfate mixture with potassium sulfide k2sx PubChem CID: 56842985 IUPAC Name: tetrapotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;sulfanide SMILES: [S--].[K+].[K+].[K+].[K+].[O-]S([O-])(=O)=S
2
Potassium Sulfide, Supelco™
CAS: 39365-88-3