accessibility menu, dialog, popup

UserName

CAS RN 39330-74-0

Er

CAS RN 39330-74-0

IUPAC Name: erbium(3+) tris(cyclopenta-1,3-diene)
Synonyms: erbium(3+) tris(cyclopentadiene)
Molecular Weight (g/mol): 365.57
Molecular Formula: C15H18Er
InChi Key: WEZXZKINGCWZRM-UHFFFAOYSA-N
SMILES: [Er+3].C1C=CC=C1.C1C=CC=C1.C1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results