accessibility menu, dialog, popup

UserName

NaOOCl

CAS RN 3926-62-3

IUPAC Name: sodium 2-chloroacetate
Synonyms: sodium chloroacetate
Molecular Weight (g/mol): 116.48
Molecular Formula: C2H2ClNaO2
InChi Key: FDRCDNZGSXJAFP-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CCl
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)