accessibility menu, dialog, popup

UserName

HONHNOHNHNOHNNHOONHOOHNO

CAS RN 39236-46-9

IUPAC Name: 3-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1-[({[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoyl}amino)methyl]urea
Synonyms: imidazolidinyl urea germall 115 imidurea
Molecular Weight (g/mol): 388.3
Molecular Formula: C11H16N8O8
InChi Key: ZCTXEAQXZGPWFG-UHFFFAOYNA-N
SMILES: OCN1C(NC(=O)NCNC(=O)NC2N(CO)C(=O)NC2=O)C(=O)NC1=O
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (5)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)