accessibility menu, dialog, popup

UserName

FNCl

CAS RN 391-77-5

IUPAC Name: 4-chloro-6-fluoroquinoline
Synonyms: 4-chloro-6-fluoroquinoline
Molecular Weight (g/mol): 181.59
Molecular Formula: C9H5ClFN
InChi Key: CKTQPWIDUQGUGG-UHFFFAOYSA-N
SMILES: FC1=CC=C2N=CC=C(Cl)C2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results