CAS RN 390-64-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Prenylamine, TRC
CAS: 390-64-7 Molecular Formula: C24 H27 N Molecular Weight (g/mol): 329.48 Synonym: Benzenepropanamine, N-(1-methyl-2-phenylethyl)-γ-phenyl-,Phenethylamine, N-(3,3-diphenylpropyl)-α-methyl- (6CI,8CI),N-(1-Methyl-2-phenylethyl)-γ-phenylbenzenepropanamine,B 436,DL-Prenylamine,Elecor,N-(3,3-Diphenylpropyl)-α-methylphenethylamine,N-(3,3-Diphenylpropyl)-α-methylphenylethylamine,Prenylamine,Prenylamine (vasodilator),SAN 13-194,Sandoz 13-194,Segontin IUPAC Name: 3,3-diphenyl-N-(1-phenylpropan-2-yl)propan-1-amine SMILES: CC(Cc1ccccc1)NCCC(c2ccccc2)c3ccccc3