accessibility menu, dialog, popup

UserName

NH3CH3CCH3CH3BrBrBr

CAS RN 38932-80-8

IUPAC Name: N,N,N-trimethylanilinium
Synonyms: tetrabutylammonium tribromide
Molecular Weight (g/mol): 136.22
Molecular Formula: C9H14N
InChi Key: ZNEOHLHCKGUAEB-UHFFFAOYSA-N
SMILES: C[N+](C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)