accessibility menu, dialog, popup

UserName

CAS RN 3888-65-1

IUPAC Name: 4-(4-fluorophenyl)piperidin-4-ol
Synonyms: 4-(4-fluorophenyl)piperidin-4-ol
Molecular Weight (g/mol): 195.24
Molecular Formula: C11H14FNO
InChi Key: QXWRXWPNHLIZBV-UHFFFAOYSA-N
SMILES: OC1(CCNCC1)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results