accessibility menu, dialog, popup

UserName

CAS RN 38869-24-8

CH3OOCs

CAS RN 38869-24-8

IUPAC Name: caesium(1+) propanoate
Synonyms: caesium(1+) propionate
Molecular Weight (g/mol): 205.98
Molecular Formula: C3H5CsO2
InChi Key: YBZSHUAKOJGWRT-UHFFFAOYSA-M
SMILES: [Cs+].CCC([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results