CAS RN 3886-69-9
(R)-(+)-1-Phenylethylamine, 99+%, ee 99+%
CAS: 3886-69-9 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
D(+)-alpha-Methylbenzylamine, 99+%, (99% ee)
CAS: 3886-69-9 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.18 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(R)-(+)-1-Phenylethylamine, 99+%
CAS: 3886-69-9 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(R)-(+)-1-Phenylethylamine, 98%
CAS: 3886-69-9 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(R)-(+)-1-Phenylethylamine, Sigma-Aldrich™
CAS: 3886-69-9 Grade: Research Molecular Formula: C6H5CH(CH3)NH2 Molecular Weight (g/mol): 121.18
(R)-(+)-alpha-Methylbenzylamine, TRC
CAS: 3886-69-9 Molecular Formula: C8 H11 N Molecular Weight (g/mol): 121.18 Synonym: Benzenemethanamine, α-methyl-, (αR)-,Benzenemethanamine, α-methyl-, (R)-,Benzylamine, α-methyl-, (R)-(+)- (8CI),Benzylamine, α-methyl-, d- (4CI),(αR)-α-Methylbenzenemethanamine,(+)-(R)-1-Phenethylamine,(+)-(R)-1-Phenylethylamine,(+)-(R)-α-Methylbenzylamine,(+)-(R)-α-Phenylethylamine,(+)-1-Amino-1-phenylethane,(+)-1-Phenethylamine,(+)-1-Phenylethan-1-amine,(+)-1-Phenylethylamine,(+)-Phenylethylamine,(+)-[(1R)-1-Phenylethyl]amine,(+)-α-Methylbenzenemethanamine,(+)-α-Methylbenzylamine,(+)-α-Phenethylamine,(+)-α-Phenylethylamine,(1R)-1-Phenyl-1-ethanamine,(1R)-1-Phenylethan-1-amine,(1R)-1-Phenylethanamine,(1R)-1-Phenylethanamine,(1R)-1-Phenylethylamine,(1R)-Phenylethylamine,(R)-(+)-1-Methylbenzylamine,(R)-(+)-1-Phenethylamine,(R)-(+)-1-Phenylethanamine,(R)-(+)-1-Phenylethylamine,(R)-(+)-Alpha-methylbenzylamine,(R)-(+)-Phenethylamine,(R)-(+)-α-Methylbenzylamine,(R)-(+)-α-Phenethylamine,(R)-(+)-α-Phenylethylamine,(R)-1-Amino-1-phenylethane,(R)-1-Aminoethylbenzene,(R)-1-Phenethylamine,(R)-1-Phenyl-1-ethanamine,(R)-1-Phenylethanamine,(R)-1-Phenylethylamine,(R)-Alpha-methylbenzylamine,(R)-Methylbenzylamine,(R)-Phenethylamine,(R)-Phenylethylamine,(R)-Phenylethylamine,(R)-α-Aminoethylbenzene,(R)-α-Methylbenzenemethanamine,(R)-α-Methylbenzylamine,(R)-α-Phenethylamine,(R)-α-Phenylethylamine,D-(+)-1-Phenylethylamine,D-(+)-α-Methylbenzylamine,D-(+)-α-Phenylethylamine,D-1-Phenylethylamine,D-Phenethylamine,D-α-Methylbenzylamine,D-α-Phenethylamine,R-(+)-Methylbenzylamine,R-(+)-Phenylethylamine,d-1-Phenylethylamine,Levetiracetam Imp. E (EP) IUPAC Name: (1R)-1-phenylethanamine SMILES: C[C@@H](N)c1ccccc1
(R)-(+)-Alpha-Methylbenzylamine (>85%), TRC
CAS: 3886-69-9 Molecular Formula: C8 H11 N Molecular Weight (g/mol): 121.18 Synonym: Benzenemethanamine, α-methyl-, (αR)-,Benzenemethanamine, α-methyl-, (R)-,Benzylamine, α-methyl-, (R)-(+)- (8CI),Benzylamine, α-methyl-, d- (4CI),(αR)-α-Methylbenzenemethanamine,(+)-(R)-1-Phenethylamine,(+)-(R)-1-Phenylethylamine,(+)-(R)-α-Methylbenzylamine,(+)-(R)-α-Phenylethylamine,(+)-1-Amino-1-phenylethane,(+)-1-Phenethylamine,(+)-1-Phenylethan-1-amine,(+)-1-Phenylethylamine,(+)-Phenylethylamine,(+)-[(1R)-1-Phenylethyl]amine,(+)-α-Methylbenzenemethanamine,(+)-α-Methylbenzylamine,(+)-α-Phenethylamine,(+)-α-Phenylethylamine,(1R)-1-Phenyl-1-ethanamine,(1R)-1-Phenylethan-1-amine,(1R)-1-Phenylethanamine,(1R)-1-Phenylethanamine,(1R)-1-Phenylethylamine,(1R)-Phenylethylamine,(R)-(+)-1-Methylbenzylamine,(R)-(+)-1-Phenethylamine,(R)-(+)-1-Phenylethanamine,(R)-(+)-1-Phenylethylamine,(R)-(+)-Alpha-methylbenzylamine,(R)-(+)-Phenethylamine,(R)-(+)-α-Methylbenzylamine,(R)-(+)-α-Phenethylamine,(R)-(+)-α-Phenylethylamine,(R)-1-Amino-1-phenylethane,(R)-1-Aminoethylbenzene,(R)-1-Phenethylamine,(R)-1-Phenyl-1-ethanamine,(R)-1-Phenylethanamine,(R)-1-Phenylethylamine,(R)-Alpha-methylbenzylamine,(R)-Methylbenzylamine,(R)-Phenethylamine,(R)-Phenylethylamine,(R)-Phenylethylamine,(R)-α-Aminoethylbenzene,(R)-α-Methylbenzenemethanamine,(R)-α-Methylbenzylamine,(R)-α-Phenethylamine,(R)-α-Phenylethylamine,D-(+)-1-Phenylethylamine,D-(+)-α-Methylbenzylamine,D-(+)-α-Phenylethylamine,D-1-Phenylethylamine,D-Phenethylamine,D-α-Methylbenzylamine,D-α-Phenethylamine,R-(+)-Methylbenzylamine,R-(+)-Phenylethylamine,d-1-Phenylethylamine,Levetiracetam Imp. E (EP) IUPAC Name: (1R)-1-phenylethanamine SMILES: C[C@@H](N)c1ccccc1
(1R)-1-Phenylethan-1-amine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.