accessibility menu, dialog, popup

UserName

CAS RN 388088-75-3

H3CNNSOClH3C

CAS RN 388088-75-3

IUPAC Name: 1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carbonyl chloride
Synonyms: 1,3-dimethylthieno[2,3-c]pyrazole-5-carbonyl chloride
Molecular Weight (g/mol): 214.67
Molecular Formula: C8H7ClN2OS
InChi Key: BQOTYHLFKDBAGL-UHFFFAOYSA-N
SMILES: CN1N=C(C)C2=C1SC(=C2)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results